Order Entry
ContactUsLinkComponent
Human Granulysin ELISA Kit, Rockland Immunochemicals
Human Granulysin ELISA Kit, Rockland Immunochemicals
Catalog #: 76235-976
CAS Number:  
Human Granulysin ELISA Kit, Rockland Immunochemicals
Catalog #: 76235-976
CAS Number:  

Specifications

  • Reactivity:
    Human
  • Description:
    Human Granulysin ELISA Kit
  • Size:
    1 Kit
  • Shipping temperature:
    Wet Ice
  • Cat. no.:
    76235-976

Specifications

About this item

Human Granulysin AccuSignal ELISA Kit

  • Natural and recombinant human Granulysin.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: M1-L145

Granulysin is a substance released by cytotoxic T cells(CD8) when they are attached to infected body cells. The product of this gene is a member of the saposin-like protein(SAPLIP) family. It is mapped to 2p11.2. Granulysin functions to create holes in the target cell membrane and destroy it. It is able to induce apoptosis in target cells and also has antimicrobial action. This gene is expressed in cytolytic granules with perforin, a pore forming protein, and granzymes that are also involved in cytolysis. In addition to it, Granulysin is broadly antimicrobial, killing microbes that cause, for example, tuberculosis and malaria, and can destroy some tumors. A series of peptides generated from the amino acid sequence of Granulysin are potential antibiotics. It has been found that secretory Granulysin is a key molecule responsible for the disseminated keratinocyte death in SJS/TEN.

Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 2000pg/ml, 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml human Granulysin standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each human Granulysin standard solution and each sample be measured in duplicate.

Customers Who Bought This Also Bought