Specifications
- Reactivity:Human
- Target protein:IL-18R1
- Description:Human IL-18R1 ELISA Kit
- Size:1 Kit
- Shipping temperature:Wet Ice
- Cat. no.:76235-952
Specifications
About this item
Human Interleukin-18R1 AccuSignal ELISA Kit
- Natural and recombinant human IL-18R1.
- There is no detectable cross-reactivity with other relevant proteins.
- Expression system for standard: NSO; Immunogen sequence: C22-R329
The interleukin-18 receptor 1(IL-18R1), also known as CDw218a(cluster of differentiation w218a) or IL18RA, is an interleukin receptor of the immunoglobulin superfamily. The protein encoded by this gene is a cytokine receptor that belongs to the interleukin 1 receptor family. This gene along with four other members of the interleukin 1 receptor family, including IL1R2, IL1R1, ILRL2(IL-1Rrp2), and IL1RL1(T1/ST2), form a gene cluster on chromosome 2q. This receptor specifically binds interleukin 18(IL18), and is essential for IL18 mediated signal transduction. IFN-alpha and IL12 are reported to induce the expression of this receptor in NK and T cells.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 2000pg/ml, 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml human IL-18R1 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each human IL-18R1 standard solution and each sample be measured in duplicate.