Specifications
- Reactivity:Human
- Description:Human Tetranectin - CLEC3B ELISA Kit
- Size:1 Kit
- Shipping temperature:Wet Ice
- Cat. no.:76235-948
Specifications
About this item
Human Tetranectin - CLEC3B AccuSignal ELISA Kit
- Natural and recombinant human Tetranectin.
- There is no detectable cross-reactivity with other relevant proteins.
- Expression system for standard: NSO; Immunogen sequence: E22-V202
Tetranectin, also called TNA, is a protein that in humans is encoded by the CLEC3B gene. It is mapped to 3p21.31. Tetranectin, a tetrameric protein isolated from human plasma, has 4 identical and noncovalently bound polypeptide chains, each of 181 amino acid residues. It has a specific binding affinity for sulfated polysaccharides and the kringle 4 of plasminogen. The plasma concentration of tetranectin is reduced in patients with various malignancies. Tetranectin is a plasminogen-binding protein that is induced during the mineralization phase of osteogenesis. Thus, tetranectin is a candidate gene for human disorders affecting bone and connective tissue.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 20,000pg/ml, 10,000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml human Tetranectin standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or tissue Homogenates to each empty well. We recommend that each human Tetranectin standard solution and each sample is measured in duplicate.