Specifications
- Reactivity:Human
- Description:Human Lumican ELISA Kit
- Size:1 Kit
- Shipping temperature:Wet Ice
- Cat. no.:76237-462
Specifications
About this item
Human Lumican AccuSignal ELISA Kit
- Natural and recombinant human Lumican.
- There is no detectable cross-reactivity with other relevant proteins.
- Expression system for standard: NSO; Immunogen sequence: Q19-N338
Lumican, also known as LUM, is a protein which in humans is encoded by the LUM gene. This gene is a member of the small interstitial proteoglycan gene(SIPG) family, and it is a keratan sulfate proteoglycan which presents large quantities in the corneal stroma and in interstitial collagenous matrices of the heart, aorta, skeletal muscle, skin, and intervertebral discs. Lumican is mapped to 12q21.33, it interacts with collagen and limits growth of fibrils in diameter. In the cornea, Lumican not only interacts with collagen molecules to limit fibril growth, but also plays a critical role in the regular spacing of fibrils and acquisition of corneal transparency by virtue of its keratan sulfate-containing glycosaminoglycan side chains LDC.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 10,000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml, 156pg/ml human Lumican standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin) to each empty well. It is recommended that each human Lumican standard solution and each sample be measured in duplicate.