Specifications
- Reactivity:Human
- Description:Human SPARC ELISA Kit
- Size:1 Kit
- Shipping temperature:Wet Ice
- Cat. no.:76235-894
Specifications
About this item
Human SPARC AccuSignal ELISA Kit
- Natural and recombinant human SPARC.
- There is no detectable cross-reactivity with other relevant proteins.
- Expression system for standard: NSO; Immunogen sequence: A18-I303
SPARC(secreted protein acidic and rich in cysteine), also known as Osteonectin, is a protein that in humans is encoded by the SPARC gene. The human SPARC gene is 26.5 kb long, and contains 10 exons and 9 introns and is located on chromosome 5q31-q33. SPARC is a glycoprotein of 40 kD. SPARC is an acidic, cysteine-rich glycoprotein consisting of a single polypeptide chain that can be broken into 4 domains: 1) an Ca++ binding domains near the glutamic acidic-rich region at the amino terminus(domain I), 2) a cysteine- rich(domain II), 3) a hydrophilic region(domain III) and 4) an EF hand motif at the carboxy terminus region(domain IV). Osteonectin is a glycoprotein in the bone that binds sodium. It is secreted by osteoblasts during bone formation, initiating mineralization and promoting mineral crystal formation. Osteonectin also shows affinity for collagen in addition to bone mineral calcium. A correlation between osteonectin over expression and ampullary cancers and chronic pancreatitis has been found.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 50ng/ml, 25ng/ml, 12.5ng/ml, 6.25ng/ml, 3.12ng/ml, 1.56ng/ml, 0.78ng/ml human SPARC standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or human milk to each empty well. We recommend that each human SPARC standard solution and each sample is measured in duplicate.