Specifications
- Reactivity:Human
- Description:Human S100A12 ELISA Kit
- Size:1 Kit
- Shipping temperature:Wet Ice
- Cat. no.:76235-824
Specifications
About this item
Human S100A12 AccuSignal ELISA Kit
- Natural and recombinant human S100A12.
- There is no detectable cross-reactivity with other relevant proteins.
- Expression system for standard: E.coli; Immunogen sequence: M1-E92
S100 calcium-binding protein A12 (S100A12) is a protein that in humans is encoded by the S100A12 gene. It is also known as calgranulin C. By direct R-banding FISH, the S100A12 gene is mapped to chromosome 1q21.2-q22. The protein encoded by this gene is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs. S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells, and involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. S100 genes include at least 13 members which are located as a cluster on chromosome 1q21. And this protein is proposed to be involved in specific calcium-dependent signal transduction pathways and its regulatory effect on cytoskeletal components may modulate various neutrophil activities.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 20,000pg/ml, 10,000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml human S100A12 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma( heparin, EDTA) to each empty well. It is recommended that each human S100A12 standard solution and each sample be measured in duplicate.