Specifications
- Assay Type:Sandwich
- Reactivity:Mouse
- Description:Mouse TLR2 ELISA Kit
- Size:1 Kit
- Shipping temperature:Wet Ice
- Cat. no.:76235-810
Specifications
About this item
Mouse TLR2 AccuSignal ELISA Kit
- Natural and recombinant mouse TLR2.
- There is no detectable cross-reactivity with other relevant proteins.
- Expression system for standard: NSO; Immunogen sequence: Q25-L590
Toll-like receptor 2 also known as TLR-2 is a protein that in humans is encoded by the TLR2 gene. TLR2 has also been designated as CD282(cluster of differentiation 282). TLR-2 plays a role in the immune system. TLR-2 is a membrane protein, a receptor, which is expressed on the surface of certain cells and recognizes foreign substances and passes on appropriate signals to the cells of the immune system. TLR2 expresses as 4- and 4.4-kb mRNAs in heart, brain, and muscle.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 10,000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml, 156pg/ml mouse TLR2 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each mouse TLR2 standard solution and each sample be measured in duplicate.