Order Entry
ContactUsLinkComponent
Human CCL14 - HCC-1 ELISA Kit, Rockland Immunochemicals
Human CCL14 - HCC-1 ELISA Kit, Rockland Immunochemicals
Catalog #: 76237-452
CAS Number:  
Human CCL14 - HCC-1 ELISA Kit, Rockland Immunochemicals
Catalog #: 76237-452
CAS Number:  

Specifications

  • Reactivity:
    Human
  • Description:
    Human CCL14 - HCC-1 ELISA Kit
  • Size:
    1 Kit
  • Shipping temperature:
    Wet Ice
  • Cat. no.:
    76237-452

Specifications

About this item

Human CCL14 - HCC-1 AccuSignal ELISA Kit

  • Natural and recombinant human CCL14.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence: T20-N93

Chemokine(C-C motif) ligand 14(CCL14) is a small cytokine belonging to the CC chemokine family. It is also commonly known as HCC-1. It is produced as a protein precursor that is processed to generate a mature active protein containing 74 amino acids that and is 46% identical in amino acid composition to CCL3 and CCL4. This chemokine is expressed in various tissues including spleen, bone marrow, liver, muscle, and gut. CCL14 activates monocytes, but does not induce their chemotaxis. In addition, HCC-1 enhanced the proliferation of CD34+ myeloid progenitor cells. It was as effective as MIP-1 alpha, but about 100-fold less potent. Human CCL14 is located on chromosome 17 within a cluster of other chemokines belonging to the CC family.

Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.3pg/ml, 15.6pg/ml human CCL14 standard solutions into the pre-coated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. We recommend that each human CCL14 standard solution and each sample is measured in duplicate.

Customers Who Bought This Also Bought