Specifications
- Reactivity:Human
- Description:Human ALCAM ELISA Kit
- Size:1 Kit
- Shipping temperature:Wet Ice
- Cat. no.:76235-764
Specifications
About this item
Human ALCAM AccuSignal ELISA Kit
- Natural and recombinant human ALCAM.
- There is no detectable cross-reactivity with other relevant proteins.
- Expression system for standard: NSO; Immunogen sequence: W28-A526
CD166 antigen is a 100-105 kD typeI transmembrane glycoprotein that is a member of the immunoglobulin superfamily of proteins. In humans it is encoded by the ALCAM gene. The gene was mapped to human chromosome 3q13.1-q13.2 by fluorescence in situ hybridization of cDNA probes to metaphase chromosomes. It is also called CD166(cluster of differentiation 166), MEMD, SC-1/DM-GRASP/BEN in the chicken, and KG-CAM in the rat. It is expressed on activated T cells, activated monocytes, epithelial cells, fibroblasts, neurons, melanoma cells, and also in sweat and sebaceous glands. CD166 protein expression is reported to be upregulated in a cell line deriving from a metastasizing melanoma. CD166 plays an important role in mediating adhesion interactions between thymic epithelial cells and CD6+ cells during intrathymic T cell development. Recently, CD166 has also been used as a potential cancer stem cell marker.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 20000pg/ml, 10000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml human ALCAM standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma (heparin, EDTA) to each empty well. We recommend that each human ALCAM standard solution and each sample is measured in duplicate.