Specifications
- Reactivity:Mouse
- Target protein:IL-12 (p40)
- Description:Mouse IL-12(p40) ELISA Kit
- Size:1 Kit
- Shipping temperature:Wet Ice
- Cat. no.:76237-422
Specifications
About this item
Mouse Interleukin-12(p40) AccuSignal ELISA Kit
- Natural and recombinant mouse IL-12(p40).
- There is no detectable cross-reactivity with other relevant proteins.
- Expression system for standard: sf21; Immunogen sequence: M23-S419
Interleukin-12(IL12, formerly NKSF, for natural killer cell stimulatory factor, or CLMF, for cytotoxic lymphocyte maturation factor) is a novel cytokine cloned from B-cell lines. IL-12 is a heterodimeric molecule composed of p35 and p40 subunits.1 The larger 40-kDa subunit(p40) is a member of the cytokine receptor family, and the smaller 35-kDa subunit(p35) is related to IL6 and GCSF.2 Both IL-12 p40(-/-) and p35(-/-) mice fail to produce IL-12 p70 heterodimer.3 Interleukin(IL)-12 has been cloned on the basis of its ability to activate natural killer(NK) cells and promote the development of cytolytic T cells. With further understanding of its activities, IL-12 has emerged as an important cytokine, affecting both immune and hematologic functions. It has been shown to be necessary for the T cell independent induction of interferon(IFN)-gamma, critical for the initial suppression of bacterial and parasitic infection; for the development of a Th1 response, critical for effective host defense against intracellular pathogens; and for the activation of differentiated T lymphocytes of both CD4+ and CD8+ phenotype.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml, 15.6pg/ml mouse IL-12(p40) standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma(heparin, EDTA, citrate) to each empty well. It is recommended that each mouse IL-12(p40) standard solution and each sample be measured in duplicate.