Specifications
- Reactivity:Human
- Target protein:IL-33
- Description:Human IL-33 ELISA Kit
- Size:1 Kit
- Shipping temperature:Wet Ice
- Cat. no.:76237-418
Specifications
About this item
Human Interleukin-33 AccuSignal ELISA Kit
- Natural and recombinant human IL-33.
- There is no detectable cross-reactivity with other relevant proteins.
- Expression system for standard: E.coli; Immunogen sequence: S112-T270
Interleukin 33(IL-33) is a cytokine belonging to the IL-1 superfamily. The IL33 gene maps to chromosome 9p24.1 by using of genomic database analysis. Recombinant mature human IL33 bound to ST2. The induction of type 2 cytokines by IL-33 in vivo is believed to induce the severe pathological changes observed in mucosal organs following administration of IL-33. IL33, an alarmin released from necrotic cells, is necessary for potent CD8 + T cell(CTL) responses to replicating, prototypic RNA, and DNA viruses in mice. IL33 prevented the downregulation of CXCR2 and inhibition of chemotaxis induced by activation of TLR4, and found that IL33 reverses the TLR4-induced reduction of CXCR2 expression via the inhibition of expression of GRK2.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml, 15.6pg/ml human IL-33 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum, plasma(heparin, EDTA) or cell lysates to each empty well. It is recommended that each human IL-33 standard solution and each sample be measured in duplicate.