Order Entry
ContactUsLinkComponent
Human TLR2 ELISA Kit, Rockland Immunochemicals
Human TLR2 ELISA Kit, Rockland Immunochemicals
Catalog #: 76237-388
CAS Number:  
Human TLR2 ELISA Kit, Rockland Immunochemicals
Catalog #: 76237-388
CAS Number:  

Specifications

  • Reactivity:
    Human
  • Description:
    Human TLR2 ELISA Kit
  • Size:
    1 Kit
  • Shipping temperature:
    Wet Ice
  • Cat. no.:
    76237-388

Specifications

About this item

Human TLR2 AccuSignal ELISA Kit

  • Natural and recombinant human TLR2.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: E21-L590

Toll-like receptor 2 also known as TLR-2 is a protein that in humans is encoded by the TLR2 gene. TLR2 has also been designated as CD282(cluster of differentiation 282). TLR-2 plays a role in the immune system. TLR-2 is a membrane protein, a receptor, which is expressed on the surface of certain cells and recognizes foreign substances and passes on appropriate signals to the cells of the immune system. TLR2 expresses as 4- and 4.4-kb mRNAs in heart, brain, and muscle..

Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 10000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml, 156pg/ml human TLR2 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin , EDTA) to each empty well. It is recommended that each human TLR2 standard solution and each sample be measured in duplicate.

Customers Who Bought This Also Bought