Specifications
- Reactivity:Human
- Description:Human P53 ELISA Kit
- Size:1 Kit
- Shipping temperature:Wet Ice
- Cat. no.:76235-668
Specifications
About this item
Human P53 AccuSignal ELISA Kit
- Natural and recombinant human P53.
- There is no detectable cross-reactivity with other relevant proteins.
- Expression system for standard: NSO; Immunogen sequence: M1-D393
P53(also known as protein 53 or tumor protein 53), is a tumor suppressor protein that in humans is encoded by the TP53 gene. The human P53 gene is mapped to chromosome 17. Human p53 is 393 amino acids long and has seven domains. It runs as a 53-kilodalton(kDa) protein on SDS-PAGE. The pattern of p53 splicing was specific for brain areas and for individuals. And human kidney and heart expressed only full-length p53. It has played a vital role in conserving stability by preventing genome mutation.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 10000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml, 156pg/ml human P53 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell lysates to each empty well. It is recommended that each human P53 standard solution and each sample be measured in duplicate.