Specifications
- Assay Type:Sandwich
- Reactivity:Mouse
- Description:Mouse KIM1 ELISA Kit
- Size:1 Kit
- Shipping temperature:Wet Ice
- Cat. no.:76235-654
Specifications
About this item
Mouse KIM1 AccuSignal ELISA Kit
- Natural and recombinant mouse KIM1.
- There is no detectable cross-reactivity with other relevant proteins.
- Expression system for standard: NSO; Immunogen sequence: Y22-T212
KIM1(TIM-1), also known as Hepatitis A virus cellular receptor 1, is a protein that in Mouses is encoded by the HAVCR1 gene.[1][2][3] Infection of canine osteogenic sarcoma cells expressing HAVCR1 with HAV led Feigelstock et al.(1998) to conclude that the protein is indeed a receptor for the virus. Immunofluorescence microscopy demonstrated internalization of HAV by dog cells expressing HAVCR1. Using a monoclonal antibody to mouse Tim1, Umetsu et al.(2005) showed that Tim1 was expressed after activation of naive T cells and on T cells differentiated in Th2-polarizing conditions. By homology of synteny with the mouse Tim1 gene and database analysis, McIntire et al.(2001) mapped the HAVCR1 gene to 5q33.2. The standard used in this kit is recombinant mouse KIM-1, constituting from Y22-T212 amino acids, and 191amino acids with the molecular mass of 21.8KDa.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 2000pg/ml, 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml mouse KIM1 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, tissue homogenates, serum, plasma(heparin, EDTA) or urine to each empty well. It is recommended that each mouse KIM1 standard solution and each sample be measured in duplicate.