Specifications
- Assay Type:Sandwich
- Reactivity:Mouse
- Description:Mouse PECAM-1 - CD31 ELISA Kit
- Size:1 Kit
- Shipping temperature:Wet Ice
- Cat. no.:76235-652
Specifications
About this item
Mouse PECAM-1 - CD31 AccuSignal ELISA Kit
- Natural and recombinant mouse PECAM-1.
- There is no detectable cross-reactivity with other relevant proteins.
- Expression system for standard: NSO; Immunogen sequence: E18-K590
PECAM-1 is an important target GP in DITP.1 Platelet/endothelial cell adhesion molecule-1(PECAM-1) is a 130-kD member of the Ig gene superfamily that is expressed on the surface of circulating platelets, monocytes, neutrophils, and selective T-cell subsets.2 Platelet endothelial cell adhesion molecule-1(PECAM-1/CD31) is a member of the immunoglobulin(Ig) superfamily that has distinctive features of an immunoreceptor based upon its genomic structure and the presence of intrinsic immunoreceptor tyrosine inhibitory motifs(ITIMs) in its ligand binding polypeptide.3 PECAM-1 has recently been shown to contain functional immunoreceptor tyrosine-based inhibitory motifs(ITIMs) within its cytoplasmic domain, and co-ligation of PECAM-1 with the T-cell antigen receptor(TCR) results in tyrosine phosphorylation of PECAM-1, recruitment of Src homology 2 domain-containing protein tyrosine phosphatase-2(SHP-2), and attenuation of TCR-mediated cellular signaling.4 The standard used in this kit is the product of gene recombinant expression(extracellular part), consisting of 563 amino acids(E28-K590) with the molecular weight of 65.5KDa. After glycosylating, its molecular weight changes to 91-100KDa.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 10000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml, 156pg/ml mouse PECAM-1 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each mouse PECAM-1 standard solution and each sample be measured in duplicate.