Order Entry
ContactUsLinkComponent
Mouse Lipocalin-2 - NGAL ELISA Kit, Rockland Immunochemicals
Mouse Lipocalin-2 - NGAL ELISA Kit, Rockland Immunochemicals
Catalog #: 76236-918
CAS Number:  
Mouse Lipocalin-2 - NGAL ELISA Kit, Rockland Immunochemicals
Catalog #: 76236-918
CAS Number:  

Specifications

  • Assay Type:
    Sandwich
  • Reactivity:
    Mouse
  • Target protein:
    Lipocalin-2/NGAL
  • Description:
    Mouse Lipocalin-2 - NGAL ELISA Kit
  • Size:
    1 Kit
  • Shipping temperature:
    Wet Ice
  • Cat. no.:
    76236-918

Specifications

About this item

Mouse Lipocalin-2 - NGAL AccuSignal ELISA Kit

  • Natural and recombinant mouse NGAL.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: Q21-N200

Lipocalin-2(LCN2), also known as NGAL, is a protein associated with neutrophil gelatinase.1 The LCN2 gene is located at 9q34 and contains 7 exons.2 The 25-kD LCN2 protein is believed to bind small lipophilic substances such as bacteria-derived lipopolysaccharide(LPS) and formylpeptides and may function as a modulator of inflammation. NGAL tightly binds bacterial catecholate-type ferric siderophores through a cyclically permuted, hybrid electrostatic/cation-pi interaction and is a potent bacteriostatic agent in iron-limiting conditions.3 The primary LCN2 transcript is 3,696 nucleotides long, and the processed transcript is 809 nucleotides long.4 LCN2 expression in adult bone marrow, uterus, prostate, salivary gland, stomach, appendix, colon, trachea, and lung, and in fetal spleen and lung. The standard product used in this kit is recombinant mouse NGAL, consisting of 179 amino acids with the molecular mass of 22KDa.

Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 313pg/ml, 156pg/ml, 78pg/ml mouse NGAL standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum, plasma(heparin) or urine to each empty well. It is recommended that each mouse NGAL standard solution and each sample be measured in duplicate.

Customers Who Bought This Also Bought