Specifications
- Reactivity:Human
- Description:Human MPO ELISA Kit
- Size:1 Kit
- Shipping temperature:Wet Ice
- Cat. no.:76235-642
Specifications
About this item
Human MPO AccuSignal ELISA Kit
- Natural and recombinant human MPO.
- There is no detectable cross-reactivity with other relevant proteins.
- Expression system for standard: NSO; Immunogen sequence: A49-S745
Myeloperoxidase(MPO) is a mammalian phagocyte hemoprotein thought to primarily mediate host defense reactions. It is abundantly expressed in neutrophils and secreted during their activation. Myeloperoxidase is part of the host defense system of human polymorphonuclear leukocytes, responsible for microbicidal activity against a wide range of organisms. It is located in the nucleus as well as in the cytoplasm. Intranuclear MPO may help to protect DNA against damage resulting from oxygen radicals produced during myeloid cell maturation and function. The standard product used in this kit is the product of gene recombination, consisting of 697(A49-S745) amino acids with the molecular mass of 80KDa.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 20000pg/ml, 10000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml human MPO standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, cell lysate, tissue lysate, serum, plasma(heparin, EDTA), saliva or urine to each empty well. It is recommended that each human MPO standard solution and each sample be measured in duplicate.