Order Entry
ContactUsLinkComponent
Mouse TREM-1 ELISA Kit, Rockland Immunochemicals
Mouse TREM-1 ELISA Kit, Rockland Immunochemicals
Catalog #: 76237-374
CAS Number:  
Mouse TREM-1 ELISA Kit, Rockland Immunochemicals
Catalog #: 76237-374
CAS Number:  

Specifications

  • Assay Type:
    Sandwich
  • Reactivity:
    Mouse
  • Description:
    Mouse TREM-1 ELISA Kit
  • Size:
    1 Kit
  • Shipping temperature:
    Wet Ice
  • Cat. no.:
    76237-374

Specifications

About this item

Mouse TREM-1 AccuSignal ELISA Kit

  • Natural and recombinant mouse TREM-1.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: A21-S202

Trem1, Triggering receptor expressed on myeloid cells-1, is encoded by Trem1 gene. The expression of Trem1 is in monocytes and neutrophils but not in lymphocytes, dendritic cells, or other cell types. Trem1 is a 30-kD glycoprotein that is reduced to 26 kD by deglycosylation, in agreement with the predicted molecular mass. The Trem1 gene which contains 4 exons maps to chromosome 6p21.1, within a TREM gene cluster and the mouse Trem1 gene maps to chromosome 17 in a region that shows homology of synteny to human chromosome 6. The expression of Trem1 is upregulated by stimulation with lipopolysaccharide(LPS), gram-negative bacteria, and fungi. Cross-linking of Trem1 on neutrophils induces interleukin-8(IL8) and myeloperoxidase secretion, while cross-linking on monocytes induces not only secretion of IL8 but also of monocyte chemotactic protein-1(MCP1, or SCYA2) and tumor necrosis factor(TNF); MCP1 and TNF secretion could be further upregulated by LPS-mediated priming. Trem1 engagement also induces upregulation of adhesion molecules(e.g., ITGB1) and costimulatory molecules(e.g., CD40). Trem1 is associated with DAP12(TYROBP), a molecule frequently associated with activating receptors.

Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 2000pg/ml,1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml mouse TREM-1 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum, plasma(heparin, EDTA), cell lysates or tissue homogenates to each empty well. It is recommended that each mouse TREM-1 standard solution and each sample be measured in duplicate.

Customers Who Bought This Also Bought