Specifications
- Assay Type:Sandwich
- Reactivity:Mouse
- Description:Mouse RANK ELISA Kit
- Size:1 Kit
- Shipping temperature:Wet Ice
- Cat. no.:76237-354
Specifications
About this item
Mouse RANK AccuSignal ELISA Kit
- Natural and recombinant mouse RANK.
- There is no detectable cross-reactivity with other relevant proteins.
- Expression system for standard: NSO; Immunogen sequence: Q30-P213
Receptor Activator of Nuclear Factor kappa B(RANK), also known as TRANCE Receptor, is a type I membrane protein that is expressed on the surface of osteoclasts and is involved in their activation upon ligand binding. RANK is a recently described TNF receptor family member, and its ligand, RANKL, promote survival of dendritic cells and differentiation of osteoclasts. RANK contains 383 amino acids in its intracellular domain(residues 234-616), which contain three putative TRAF-binding domains(termed I, II, and III). RANK interacts with various TRAFs through distinct motifs and activates NF-kappaB via a novel TRAF6 interaction motif, which then activates NIK, thus leading to NF-kappaB activation, whereas RANK most likely activates JNK through a TRAF2-interacting region in RANK.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 4000pg/ml, 2000pg/ml,1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml mouse RANK standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates or serum to each empty well. It is recommended that each mouse RANK standard solution and each sample be measured in duplicate.