Order Entry
ContactUsLinkComponent
Human Kallikrein-6 ELISA Kit, Rockland Immunochemicals
Human Kallikrein-6 ELISA Kit, Rockland Immunochemicals
Catalog #: 76237-616
CAS Number:  
Human Kallikrein-6 ELISA Kit, Rockland Immunochemicals
Catalog #: 76237-616
CAS Number:  

Specifications

  • Reactivity:
    Human
  • Description:
    Human Kallikrein-6 ELISA Kit
  • Size:
    1 Kit
  • Shipping temperature:
    Wet Ice
  • Cat. no.:
    76237-616

Specifications

About this item

Human Kallikrein-6 AccuSignal ELISA Kit

  • Natural and recombinant human Kallikrein-6.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: L22-K244

KLK6(Kallikrein-related peptidase 6), also called KALLIKREIN 6, NEUROSIN, PROTEASE M, ZYME or PRSS9, is a protein that in humans is encoded by the KLK6 gene. This gene is one of the fifteen kallikrein subfamily members located in a cluster on chromosome 19. The encoded enzyme is regulated by steroid hormones. Northern blot analysis revealed that the PRSS9 mRNA was expressed in several primary tumors and cell lines from mammary, prostate, and ovarian cancers, but was not detected in any metastases of these cancers. The KLK6 gene is mapped on 19q13.41. In tissue culture, the enzyme has been found to generate amyloidogenic fragments from the amyloid precursor protein, suggesting a potential for involvement in Alzheimer's disease. Upon cellular stress, neurosin was released from mitochondria to the cytosol, which resulted in the increase of degraded alpha-synuclein species. Neurosin may play a significant role in physiologic alpha-synuclein degradation and also in the pathogenesis of synucleinopathies.

Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml, 156pg/ml, 78pg/ml human Kallikrein-6 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, cell lysates, serum, plasma(heparin, EDTA) or tissue homogenates to each empty well. We recommend that each human Kallikrein-6 standard solution and each sample is measured in duplicate.

Customers Who Bought This Also Bought