Order Entry
ContactUsLinkComponent
Human Kallikrein 3 ELISA Kit, Rockland Immunochemicals
Human Kallikrein 3 ELISA Kit, Rockland Immunochemicals
Catalog #: 76237-612
CAS Number:  
Human Kallikrein 3 ELISA Kit, Rockland Immunochemicals
Catalog #: 76237-612
CAS Number:  

Specifications

  • Reactivity:
    Human
  • Description:
    Human Kallikrein 3 ELISA Kit
  • Size:
    1 Kit
  • Shipping temperature:
    Wet Ice
  • Cat. no.:
    76237-612

Specifications

About this item

Human Kallikrein 3 AccuSignal ELISA Kit

  • Natural and recombinant human Kallikrein 3.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: A18-P261

Prostate-specific antigen(PSA), also known as kallikrein III, seminin, semenogelase, gamma-seminoprotein and P-30 antigen) is a 34 kD glycoprotein manufactured almost exclusively by the prostate gland; PSA is produced for the ejaculate where it liquifies the semen in the seminal coagulum and allows sperm to swim freely.1It is also believed to be instrumental in dissolving the cervical mucous cap, allowing the entry of sperm.2It is a serine protease enzyme, the gene of which is located on the nineteenth chromosome(19q13).3

Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 20000pg/ml, 10000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml human Kallikrein 3 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each human Kallikrein 3 standard solution and each sample be measured in duplicate.

Customers Who Bought This Also Bought