Specifications
- Reactivity:Human
- Description:Human CSF3R - G-CSF R ELISA Kit
- Size:1 Kit
- Shipping temperature:Wet Ice
- Cat. no.:76235-622
Specifications
About this item
Human CSF3R - G-CSF R AccuSignal ELISA Kit
- Natural and recombinant human CSF3R.
- There is no detectable cross-reactivity with other relevant proteins.
- Expression system for standard: E.cNSO; Immunogen sequence: E25-P621
The granulocyte colony-stimulating factor receptor (G-CSF-R), also known as CD114 (Cluster of Differentiation 114), is a protein that in humans is encoded by the CSF3R gene. It is mapped to 1p35-p34.3. G-CSF-R is a cell-surface receptor for the granulocyte colony-stimulating factor (G-CSF). And the G-CSF-R is a transmembrane receptor that consists of an extracellular ligand-binding portion, a transmembrane domain, and the cytoplasmic portion that is responsible for signal transduction. In addition, GCSF-R ligand-binding is associated with dimerization of the receptor and signal transduction through proteins including Jak, Lyn, STAT, andErk1/2.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 2000pg/ml, 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml human CSF3R standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum and plasma(heparin, EDTA) to each empty well. It is recommended that each human CSF3R standard solution and each sample be measured in duplicate.