Specifications
- Reactivity:Human
- Description:Human Galectin-3 - LGALS3 ELISA Kit
- Size:1 Kit
- Shipping temperature:Wet Ice
- Cat. no.:76235-618
Specifications
About this item
Human Galectin-3 - LGALS3 AccuSignal ELISA Kit
- Natural and recombinant human Galectin-3.
- There is no detectable cross-reactivity with other relevant proteins.
- Expression system for standard: E.colI; Immunogen sequence: A2-I250
Galectin-3(GAL3), also known as LGALS3, MAC2 or GALBP, is a member of the lectin family, of which 14 mammalian galectins have been identified. Galectin-3 is encoded by a single gene, LGALS3, located on chromosome 14, locus q21–q22. It is expressed in the nucleus, cytoplasm,mitochondrion, cell surface, and extracellular space. Studies have also shown that the expression of galectin-3 is implicated in a variety of processes associated with heart failure, including myofibroblast proliferation, fibrogenesis, tissue repair, inflammation, and Ventricular remodeling. Galectin-3 is expressed in various tissues and organs, but is significantly absent in normal hepatocytes.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 10000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml, 156pg/ml human Galectin-3 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each human Galectin-3 standard solution and each sample be measured in duplicate.