Specifications
- Assay Type:Sandwich
- Reactivity:Mouse
- Description:Mouse CXCL16 ELISA Kit
- Size:1 Kit
- Shipping temperature:Wet Ice
- Cat. no.:76237-296
Specifications
About this item
Mouse CXCL16 AccuSignal ELISA Kit
- Natural and recombinant mouse CXCL16.
- There is no detectable cross-reactivity with other relevant proteins.
- Expression system for standard: E.coli; Immunogen sequence: N27-P114
Chemokine(C-X-C motif) ligand 16(CXCL16) is a small cytokine belonging to the CXC chemokine family. Larger than other chemokines(with 254 amino acids), CXCL16 is composed of a CXC chemokine domain, a mucin-like stalk, a transmembrane domain and a cytoplasmic tail containing a potential tyrosine phosphorylation site that may bind SH2. These are unusual features for a chemokine, and allow CXCL16 to be expressed as a cell surface bound molecule, as well as a soluble chemokine. CXCL16 is produced by dendritic cells found in the T cell zones of lymphoid organs, and by cells found in the red pulp of the spleen. Cells that bind and migrate in response to CXCL16 include several subsets of T cells, and natural killer T(NKT) cells. CXCL16 interacts with the chemokine receptor CXCR6, also known as Bonzo. Expression of CXCL16 is induced by the inflammatory cytokines IFN-gamma and TNF-alpha.2 The gene for human CXCL16 is located on chromosome 17p13. The standard product used in this kit is recombinant mouse CXCL16, consis...
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml, 15.6pg/ml mouse CXCL16 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each mouse CXCL16 standard solution and each sample be measured in duplicate.