Specifications
- Reactivity:Human
- Description:Human CXCL11 - I-TAC ELISA Kit
- Size:1 Kit
- Shipping temperature:Wet Ice
- Cat. no.:76235-596
Specifications
About this item
Human CXCL11 - I-TAC AccuSignal ELISA Kit
- Natural and recombinant human CXCL11.
- There is no detectable cross-reactivity with other relevant proteins.
- Expression system for standard: E.coli; Immunogen sequence: F22-F94
Chemokine(C-X-C motif) ligand 11(CXCL11) is a small cytokine belonging to the CXC chemokine family that is also called Interferon-inducible T-cell alpha chemoattractant(I-TAC) and Interferon-gamma-inducible protein 9(IP-9). By PCR analysis of somatic cell hybrids, the CXCL11 gene was mapped to chromosome 4, and using FISH, it was located to 4q21.2. The CXCL11 was strongly induced by IFNG and beta-interferon, but only weakly induced by alpha-interferon. Induction of CXCL11 required expression of STAT1. What's more, it found that monocyte chemotaxis occurs in response to recombinant CXCL11. RT-PCR analysis detected CXCL11 expression in brain tissue from a patient with AIDS dementia and a patient with multiple sclerosis but not in 2 controls.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 4000pg/ml, 2000pg/ml,1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml human CXCL11 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each human CXCL11 standard solution and each sample be measured in duplicate.