Specifications
- Assay Type:Sandwich
- Reactivity:Mouse
- Description:Mouse CXCL9 ELISA Kit
- Size:1 Kit
- Shipping temperature:Wet Ice
- Cat. no.:76235-590
Specifications
About this item
Mouse CXCL9 AccuSignal ELISA Kit
- Natural and recombinant mouse CXCL9.
- There is no detectable cross-reactivity with other relevant proteins.
- Expression system for standard: E.coli; Immunogen sequence: T22-T126
Chemokine (C-X-C motif) ligand 9 (CXCL9) is a small cytokine belonging to the CXC chemokine family that is also known as Monokine induced by gamma interferon (MIG). CXCL9 is a T-cell chemoattractant, which is induced by IFN-gamma. It is closely related to two other CXC chemokines called CXCL10 and CXCL11, whose genes are located near the gene for CXCL9 on human chromosome 4. CXCL9, CXCL10 and CXCL11 all elicit their chemotactic functions by interacting with the chemokine receptor CXCR3. The standard product used in this kit is recombinant human CXCL9, consisting of 103 amino acids with the molecular mass of 11.7KDa.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 2000pg/ml,1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml mouse CXCL9 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma(EDTA) to each empty well. It is recommended that each mouse CXCL9 standard solution and each sample be measured in duplicate.