Specifications
- Reactivity:Human
- Description:Human CXCL5 - ENA-78 ELISA Kit
- Size:1 Kit
- Shipping temperature:Wet Ice
- Cat. no.:76235-584
Specifications
About this item
Human CXCL5 - ENA-78 AccuSignal ELISA Kit
- Natural and recombinant human CXCL5.
- There is no detectable cross-reactivity with other relevant proteins.
- Expression system for standard: E.coli; Immunogen sequence: A37-N114
C-X-C motif chemokine 5 is a protein that in humans encoded by the CXCL5 gene. The protein encoded by this gene, CXCL5 is a small cytokine belonging to the CXC chemokine family that is also known as epithelial-derived neutrophil-activating peptide 78(ENA-78). It is produced following stimulation of cells with the inflammatory cytokines interleukin-1or tumor necrosis factor-alpha. Expression of CXCL5 has also been observed in eosinophils, and can be inhibited with the type II interferon IFN-gamma. This chemokine stimulates the chemotaxis of neutrophils possessing angiogenic properties. It elicits these effects by interacting with the cell surface chemokine receptor CXCR2. The gene for CXCL5 is encoded on four exons and is located on humanchromosome 4 amongst several other CXC chemokine genes. CXCL5 has been implicated in connective tissue remodeling. CXCL5 plays a role in reducing sensitivity to sunburn pain in some subjects, and is a potential target which can be utilized to understand more about pain in other inflammatory conditions like arthritis and cystitis.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 2000pg/ml,1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml human CXCL5 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA, citrate) to each empty well. It is recommended that each human CXCL5 standard solution and each sample be measured in duplicate.