Specifications
- Assay Type:Sandwich
- Reactivity:Mouse
- Description:Mouse CXCL4 - PF4 ELISA Kit
- Size:1 Kit
- Shipping temperature:Wet Ice
- Cat. no.:76237-290
Specifications
About this item
Mouse CXCL4 - PF4 AccuSignal ELISA Kit
- Natural and recombinant mouse CXCL4.
- There is no detectable cross-reactivity with other relevant proteins.
- Expression system for standard: E.coli; Immunogen sequence: V30-S105
Platelet factor 4(PF4) is a small cytokine belonging to the CXC chemokine family that is also known as chemokine(C-X-C motif) ligand 4(CXCL4). The PF4 gene was localized on map to 4q12-q13. Chemokines play fundamental roles in the development, homeostasis, and function of the immune system, and they have effects on cells of the central nervous system as well as on endothelial cells involved in angiogenesis or angiostasis. Platelet factor-4 is a 70-amino acid protein that is released from the alpha-granules of activated platelets and binds with high affinity to heparin. Its major physiologic role appears to be neutralization of heparin-like molecules on the endothelial surface of blood vessels, thereby inhibiting local antithrombin III activity and promoting coagulation. As a strong chemoattractant for neutrophils and fibroblasts, PF4 probably has a role in inflammation and wound repair.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml, 15.6pg/ml mouse CXCL4 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each mouse CXCL4 standard solution and each sample be measured in duplicate.