Specifications
- Assay Type:Sandwich
- Reactivity:Mouse
- Description:Mouse CXCL1 ELISA Kit
- Size:1 Kit
- Shipping temperature:Wet Ice
- Cat. no.:76237-282
Specifications
About this item
Mouse CXCL1 AccuSignal ELISA Kit
- Natural and recombinant mouse CXCL1.
- There is no detectable cross-reactivity with other relevant proteins.
- Expression system for standard: E.coli; Immunogen sequence: R20-K96
Chemokine(C-X-C motif) ligand 1(CXCL1) is a small cytokine belonging to the CXC chemokine family that was previously called GRO1 oncogene, GROalpha, KC, Neutrophil-activating protein 3(NAP-3) and melanoma growth stimulating activity, alpha(MSGA-alpha). In humans, this protein is encoded by the CXCL1 gene. The gene for CXCL1 is located on human chromosome 4 amongst genes for other CXC chemokines. The mature form of CXCL1 is maximally 73 amino acids long. CXCL1 is secreted by human melanoma cells, has mitogenic properties and is implicated in melanoma pathogenesis. CXCL1 is expressed by macrophages, neutrophils and epithelial cells, and has neutrophil chemoattractant activity. This chemokine elicits its effects by signaling through the chemokine receptor CXCR2.CXCL1 decreased the severity of multiple sclerosis and may offer a neuro-protective function.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml, 15.6pg/ml mouse CXCL1 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates or serum to each empty well. It is recommended that each mouse CXCL1 standard solution and each sample be measured in duplicate.