Specifications
- Reactivity:Human
- Description:Human CD26 - DPP4 ELISA Kit
- Size:1 Kit
- Shipping temperature:Wet Ice
- Cat. no.:76237-260
Specifications
About this item
Human CD26 - DPP4 AccuSignal ELISA Kit
- Natural and recombinant human CD26.
- There is no detectable cross-reactivity with other relevant proteins.
- Expression system for standard: NSO; Immunogen sequence: D34-P766
Dipeptidyl peptidase-4(DPP4), also known as adenosine deaminase complexing protein 2 or CD26(cluster of differentiation 26) is a protein that, in humans, is encoded by the DPP4 gene. By fluorescence in situ hybridization, the CD26 gene was mapped to 2q24.3. DPP4 plays a major role in glucose metabolism. It is responsible for the degradation of incretins such as GLP-1. Furthermore, it appears to work as a suppressor in the development of cancer and tumours. CD26/ DPP4 plays an important role in tumor biology, and is useful as a marker for various cancers, with its levels either on the cell surface or in the serum increased in some neoplasms and decreased in others.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 20000pg/ml, 10000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml human CD26 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum, plasma( heparin, EDTA) saliva or urine to each empty well. It is recommended that each human CD26 standard solution and each sample be measured in duplicate.