Specifications
- Reactivity:Human
- Description:Human CCL17 - TARC ELISA Kit
- Size:1 Kit
- Shipping temperature:Wet Ice
- Cat. no.:76236-878
Specifications
About this item
Human CCL17 - TARC AccuSignal ELISA Kit
- Natural and recombinant human CCL17.
- There is no detectable cross-reactivity with other relevant proteins.
- Expression system for standard: E.coli; Immunogen sequence: A24-S94
Chemokine(C-C motif) ligand 17(CCL17) is a small cytokine belonging to the CC chemokine family that is also known as thymus and activation regulated chemokine(TARC). CCL17 is expressed constitutively in thymus, but only transiently in phytohemagglutinin-stimulated peripheral blood mononuclear cells.1 This chemokine specifically binds and induces chemotaxis in T cells and elicits its effects by interacting with the chemokine receptor CCR4.1,2The gene for CCL17 is located on chromosome 16, in humans, along with other chemokines called CCL22 and CX3CL1.3,4 The standard used in this kit is recombinant human CCL17, consisting of 71 amino acids with the molecular weight of 8Kda.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 2000pg/ml,1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml human CCL17 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each human CCL17 standard solution and each sample be measured in duplicate.