Specifications
- Assay Type:Sandwich
- Reactivity:Mouse
- Description:Mouse Cystatin C ELISA Kit
- Size:1 Kit
- Shipping temperature:Wet Ice
- Cat. no.:76237-248
Specifications
About this item
Mouse Cystatin C AccuSignal ELISA Kit
- Natural and recombinant mouse Cystatin C.
- There is no detectable cross-reactivity with other relevant proteins.
- Expression system for standard: NSO; Immunogen sequence: M1-A140
Cystatin C or cystatin 3(formerly gamma trace, post-gamma-globulin or neuroendocrine basic polypeptide), a protein encoded by the CST3 gene, was originally described as a constituent of normal cerebrospinal fluid(CSF) and of urine from patients with renal failure. Cystatin 3 has a low molecular weight(approximately 13.3 kilodaltons), and it is removed from the bloodstream by glomerular filtration in the kidneys. In mouses, all cells with a nucleus(cell core containing the DNA) produce cystatin C as a chain of 120 amino acids. It is found in virtually all tissues and bodily fluids. Cystatin C, which belongs to the type II cystatin gene family, is a potent inhibitor of lysosomal proteinases(enzymes from a special subunit of the cell that break down proteins) and probably one of the most important extracellular inhibitors of cysteine proteases(it prevents the breakdown of proteins outside the cell by a specific type of protein degrading enzymes). Moreover, cystatin C is involved in network reorganization in the ...
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 20000pg/ml, 10000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml mouse Cystatin C standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum, plasma( heparin, EDTA) or urine to each empty well. It is recommended that each mouse Cystatin C standard solution and each sample be measured in duplicate.