Order Entry
ContactUsLinkComponent
Human Cystatin C ELISA Kit, Rockland Immunochemicals
Human Cystatin C ELISA Kit, Rockland Immunochemicals
Catalog #: 76237-246
CAS Number:  
Human Cystatin C ELISA Kit, Rockland Immunochemicals
Catalog #: 76237-246
CAS Number:  

Specifications

  • Reactivity:
    Human
  • Description:
    Human Cystatin C ELISA Kit
  • Size:
    1 Kit
  • Shipping temperature:
    Wet Ice
  • Cat. no.:
    76237-246

Specifications

About this item

Human Cystatin C AccuSignal ELISA Kit

  • Natural and recombinant human Cystatin C.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: M1-A146

Cystatin C or cystatin 3(formerly gamma trace, post-gamma-globulin or neuroendocrine basic polypeptide), a protein encoded by the CST3 gene, was originally described as a constituent of normal cerebrospinal fluid(CSF) and of urine from patients with renal failure.1 Cystatin 3 has a low molecular weight(approximately 13.3 kilodaltons), and it is removed from the bloodstream by glomerular filtration in the kidneys. In humans, all cells with a nucleus(cell core containing the DNA) produce cystatin C as a chain of 120 amino acids. It is found in virtually all tissues and bodily fluids. Cystatin C, which belongs to the type II cystatin gene family, is a potent inhibitor of lysosomal proteinases2(enzymes from a special subunit of the cell that break down proteins) and probably one of the most important extracellular inhibitors of cysteine proteases3(it prevents the breakdown of proteins outside the cell by a specific type of protein degrading enzymes). Moreover, cystatin C is involved in network reorganization in t...

Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 20000pg/ml, 10000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml human Cystatin C standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum, plasma( heparin, EDTA), saliva, urine or human milk to each empty well. It is recommended that each human Cystatin C standard solution and each sample be measured in duplicate.

Customers Who Bought This Also Bought