Order Entry
ContactUsLinkComponent
Human Cathepsin D ELISA Kit, Rockland Immunochemicals
Human Cathepsin D ELISA Kit, Rockland Immunochemicals
Catalog #: 76237-240
CAS Number:  
Human Cathepsin D ELISA Kit, Rockland Immunochemicals
Catalog #: 76237-240
CAS Number:  

Specifications

  • Reactivity:
    Human
  • Description:
    Human Cathepsin D ELISA Kit
  • Size:
    1 Kit
  • Shipping temperature:
    Wet Ice
  • Cat. no.:
    76237-240

Specifications

About this item

Human Cathepsin D AccuSignal ELISA Kit

  • Natural and recombinant human Cathepsin D.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: L21-L412

Cathepsin D is a protein that in humans is encoded by the CTSD gene. This proteinase is a member of the peptidase C1 family, having a specificity similar to but narrower than that of pepsin A. It is mapped to 11p15.5. The cDNA encodes a 412-amino acid protein with 20 and 44 amino acids in a pre- and prosegment, respectively. Cathepsin D is one of the lysosomal proteinases. It is ubiquitously expressed and is involved in proteolytic degradation, cell invasion, and apoptosis. Mutations in this gene are involved in the pathogenesis of several diseases, including breast cancer and possibly Alzheimer disease and it has been considered as a breast cancer tumor marker.

Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 10000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml, 156pg/ml human Cathepsin D standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each human Cathepsin D standard solution and each sample be measured in duplicate.

Customers Who Bought This Also Bought