Specifications
- Reactivity:Human
- Description:Human Cadherin-2 - N-Cadherin ELISA Kit
- Size:1 Kit
- Shipping temperature:Wet Ice
- Cat. no.:76237-234
Specifications
About this item
Human Cadherin-2 - N-Cadherin AccuSignal ELISA Kit
- Natural and recombinant human Cadherin-2.
- There is no detectable cross-reactivity with other relevant proteins.
- Expression system for standard: NSO; Immunogen sequence: D160-A724
Cadherin-2(CDH2), also known as neural cadherin(NCAD), is a protein that in humans is encoded by the CDH2 gene. It is a classical cadherin from the cadherin superfamily. This gene is mapped to 18q12.1. Cadherin-2 is expressed in the brain, skeletal and cardiac muscle. Cadherin-2 is commonly found in cancer cells and provides a mechanism for transendothelial migration. It is a calcium dependent cell-cell adhesion glycoprotein comprising five extracellular cadherin repeats, a transmembrane region and a highly conserved cytoplasmic tail. The protein functions during gastrulation and is required for establishment of left-right asymmetry. At certain central nervous system synapses, presynaptic to postsynaptic adhesion is mediated at least in part by this gene product.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 50ng/ml, 25ng/ml, 12.5ng/ml, 6.25ng/ml, 3.12ng/ml, 1.56ng/ml, 0.78ng/ml human Cadherin-2 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. We recommend that each human Cadherin-2 standard solution and each sample is measured in duplicate.