Order Entry
ContactUsLinkComponent
Rat MMP-2 ELISA Kit, Rockland Immunochemicals
Rat MMP-2 ELISA Kit, Rockland Immunochemicals
Catalog #: 76237-202
CAS Number:  
Rat MMP-2 ELISA Kit, Rockland Immunochemicals
Catalog #: 76237-202
CAS Number:  

Specifications

  • Target protein:
    MMP-2
  • Description:
    Rat MMP-2 ELISA Kit
  • Size:
    1 Kit
  • Shipping temperature:
    Wet Ice
  • Cat. no.:
    76237-202

Specifications

About this item

Rat MMP-2 AccuSignal ELISA Kit

  • Natural and recombinant rat total MMP-2.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: A30-C662

Type IV collagenase, 72-kD, is officially designated matrix metalloproteinase-2(MMP2). It is also known as gelatinase, 72-kD. MMP-2 plays an essential role in angiogenesis and arteriogenesis, two processes critical to restoration of tissue perfusion after ischemia. MMP-2 expression is increased in tissue ischemia, but the responsible mechanisms remain unknown.1 Matrix metalloproteinases(MMPs) catalyze extracellular matrix degradation. Control of their activity is a promising target for therapy of diseases characterized by abnormal connective tissue turnover. MMPs are expressed as latent proenzymes that are activated by proteolytic cleavage that triggers a conformational change in the propeptide(cysteine switch). The structure of proMMP-2 reveals how the propeptide shields the catalytic cleft and that the cysteine switch may operate through cleavage of loops essential for propeptide stability.2 The gene is localized to 16q21 using somatic cell hybrids and in situ hybridization.3 The standard product used in this kit is recombinant rat MMP-2, consisting of 662 amino acids with the molecular mass of 72KDa. The detected MMP-2 includes zymogen and active enzyme.

Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 40000pg/ml, 20,000pg/ml,10,000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml rat MMP-2 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of rat cell culture supernates, serum or plasma (heparin) to each empty well. It is recommended that each rat MMP-2 standard solution and each sample be measured in duplicate.

Customers Who Bought This Also Bought