Specifications
- Reactivity:Human
- Description:Human Prolactin ELISA Kit
- Size:1 Kit
- Shipping temperature:Wet Ice
- Cat. no.:76236-872
Specifications
About this item
Human Prolactin AccuSignal ELISA Kit
- Natural and recombinant human Prolactin.
- There is no detectable cross-reactivity with other relevant proteins.
- Expression system for standard: E.coli; Immunogen sequence: L29-C227
Prolactin, also calle PRL, is a protein that in humans is encodes by the PRL gene. This gene is mapped to 6p22.3. This gene encodes the anterior pituitary hormone prolactin. It may play a role in cell survival by suppressing apoptosis, and it is essential for lactation. Prolactin also acts in a cytokine-like manner and as an important regulator of the immune system. Prolactin has important cell cycle related functions as a growth-, differentiating- and anti-apoptotic factor. As a growth factor binding to cytokine like receptors it has also profound influence on hematopoiesis,angiogenesis and is involved in the regulation of blood clotting through several pathways.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 10000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml, 156pg/ml human Prolactin standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, citrate) to each empty well. It is recommended that each human Prolactin standard solution and each sample be measured in duplicate.