Specifications
- Assay Type:Sandwich
- Reactivity:Rat
- Target protein:TIMP-1
- Description:Rat TIMP-1 ELISA Kit
- Size:1 Kit
- Shipping temperature:Wet Ice
- Cat. no.:76236-860
Specifications
About this item
Rat TIMP-1 AccuSignal ELISA Kit
- Natural and recombinant rat TIMP-1.
- There is no detectable cross-reactivity with other relevant proteins.
- Expression system for standard: NSO; Immunogen sequence: C24-A217
The tissue inhibitor of metalloproteinases 1(TIMP1) is also called erythroid-potentiating activity(EPA). The X-linked gene for human TIMP1 is expressed in some but not all inactive X-containing somatic-cell hybrids, suggesting that this gene is either prone to reactivation or variable in its inactivation.1 Purified EPA specifically stimulates human and murine cells of the erythroid lineage, unlike murine interleukin-3(IL-3) which stimulates precursor cells from all haematopoietic lineages.2 TIMP1 is thought to play a regulatory role in connective tissues by forming inactive complexes with those metalloproteinases that are normally responsible for connective tissue turnover. The human gene encoding TIMP has been mapped to the X chromosome in the region Xp11.1-p11.4.3 The standard product used in this kit is recombinant rat TIMP-1, consisting of 194 amino acids with the molecular mass of 21.5KDa. As a result of glycosylation, the molecular mass is 32-34KDa.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 2000pg/ml, 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml rat TIMP-1 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of rat cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each rat TIMP-1 standard solution and each sample be measured in duplicate.