Specifications
- Reactivity:Human
- Target protein:EG-VEGF
- Description:Human EG-VEGF ELISA Kit
- Size:1 Kit
- Shipping temperature:Wet Ice
- Cat. no.:76237-608
Specifications
About this item
Human EG-VEGF AccuSignal ELISA Kit
- Natural and recombinant human EG-VEGF.
- There is no detectable cross-reactivity with other relevant proteins.
- Expression system for standard: NSO; Immunogen sequence: A20-F105
EG-VEGF, Endocrine glandderived vascular endothelial growth factor, is a human protein encoded by by the PROKR1 gene. Its gene is mapped to chromosome 1. The 1.4-kb cDNA encodes a protein of 105 amino acids with a well defined signal sequence. The mature protein is predicted to have 86 amino acids. Northern blot analysis demonstrated expression in testis, ovary, adrenal gland, and placenta. EG-VEGF has played a vital role in inducing proliferation, migration, and fenestration in capillary endothelial cells derived from endocrine glands. The standards of this kit are recombinant human EG-VEGF(A20-F105), with molecular weight of 10.5 KDa.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml, 15.6pg/ml human EG-VEGF standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each human EG-VEGF standard solution and each sample be measured in duplicate.