Specifications
- Assay Type:Sandwich
- Reactivity:Mouse
- Description:Mouse TNFR2 ELISA Kit
- Size:1 Kit
- Shipping temperature:Wet Ice
- Cat. no.:76236-808
Specifications
About this item
Mouse TNFR2 AccuSignal ELISA Kit
- Natural and recombinant mouse TNFR2.
- There is no detectable cross-reactivity with other relevant proteins.
- Expression system for standard: NSO; Immunogen sequence: V23-G258
Tumor necrosis factor receptor 2(TNFR2) is one of receptors of TNF. TNF has proinflammatory and immunosuppressive properties that may segregate at the level of the 2 TNF receptors(TNFRs), TNFR1 and TNFR2. The genes for TNFR1, a 55-kDa protein, and TNFR2, a 70-kDa protein, have been mapped to human chromosomes 12(12pter-cen) and 1(1pter-p32), respectively. TNFR2 was induced on glomerular endothelial cells of nephritic kidneys, and TNFR2 expression on intrinsic cells, but not leukocytes, was essential for glomerulonephritis and glomerular complement deposition. TNFR1 promotes systemic immune responses and renal T cell death, while intrinsic cell TNFR2 plays a critical role in complement-dependent tissue injury. Therefore, therapeutic blockade specifically of TNFR2 may be a promising strategy in the treatment of immune-mediated glomerulonephritis.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml, 15.6pg/ml mouse TNFR2 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates or serum to each empty well. It is recommended that each mouse TNFR2 standard solution and each sample be measured in duplicate.