Specifications
- Assay Type:Sandwich
- Reactivity:Mouse
- Target protein:TIMP-1
- Description:Mouse TIMP-1 ELISA Kit
- Size:1 Kit
- Shipping temperature:Wet Ice
- Cat. no.:76236-788
Specifications
About this item
Mouse TIMP-1 AccuSignal ELISA Kit
- Natural and recombinant mouse TIMP-1.
- There is no detectable cross-reactivity with other relevant proteins.
- Expression system for standard: NSO; Immunogen sequence: C25-R205
TIMP metallopeptidase inhibitor 1, also known as TIMP1, a tissue inhibitor of metalloproteinases, is a glycoprotein that is expressed from the several tissues of organisms. This protein is a member of the TIMP family. It was found to reside at Xp11.4-p11.1. This is the first growth factor found to be X-linked. The glycoprotein is a natural inhibitor of the matrix metalloproteinases (MMPs), a group of peptidases involved in degradation of the extracellular matrix. In addition to its inhibitory role against most of the known MMPs, the encoded protein is able to promote cell proliferation in a wide range of cell types, and may also have an anti-apoptotic function. What’s more, TIMP1 is thought to play a regulatory role in connective tissues by forming inactive complexes with those metalloproteinases that are normally responsible for connective tissue turnover.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 2000pg/ml,1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml mouse TIMP-1 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each mouse TIMP-1 standard solution and each sample be measured in duplicate.