Specifications
- Reactivity:Human
- Description:Human OPN ELISA Kit
- Size:1 Kit
- Shipping temperature:Wet Ice
- Cat. no.:76236-770
Specifications
About this item
Human OPN AccuSignal ELISA Kit
- Natural and recombinant human OPN.
- There is no detectable cross-reactivity with other relevant proteins.
- Expression system for standard: NSO; Immunogen sequence: E22-L401
Osteopontin(OPN) also called urinary stone protein, secreted phosphoprotein 1(SPP1), bone sialoprotein, and early T lymphocyte activation 1(ETA1). Osteopontin is a phosphorylated glycoprotein secreted to the mineralizing extrOPNllular matrix by osteoblasts during bone development. It is believed to facilitate the attachment of osteoblasts and osteoclasts to the extrOPNllular matrix, allowing them to perform their respective functions during osteogenesis. Osteopontin is presumably involved in stone formation as stone matrix. The deduced protein sequence reveals a 317-amino acid protein(34,982 Da) containing a 16-amino acid hydrophobic signal sequence and a 33,352-Da protein destines to undergo extensive post-translational modifications before being secreted from the cell. The gene is located on human chromosome 4. The standard product used in this kit is recombinant human OPN, consisting of 17-314 amino acid sequence with the molecular mass of 32.9KDa. As a result of glycosylation, the molecular mass is 60-65KDa.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 10000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml, 156pg/ml human OPN standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernatants, serum, plasma(heparin, EDTA), milk or urine to each empty well. It is recommended that each human OPN standard solution and each sample be measured in duplicate.