Specifications
- Reactivity:Human
- Description:Human NT-4 ELISA Kit
- Size:1 Kit
- Shipping temperature:Wet Ice
- Cat. no.:76237-592
Specifications
About this item
Human NT-4 AccuSignal ELISA Kit
- Natural and recombinant human NT-4.
- There is no detectable cross-reactivity with other relevant proteins.
- Expression system for standard: sf21; Immunogen sequence: G81-A210
Neurotrophin-4(NT-4), also known as neurotrophin-5(NT-5), is a protein that in humans is encoded by the NTF4 gene. Human NT-4 as well as a human NT-4 pseudogene colocalize to chromosome 19 band q13.3. NT-4 is a member of a family of neurotrophic factors, the neurotrophins, that control survival and differentiation of vertebrate neurons(2-4). NT-4 is a neurotrophic factor that signals predominantly through the TrkB receptor tyrosine kinase. NT4 plays a physiological role essential for hippocampus- and amygdala-dependent long-term memory and hippocampal long-lasting LTP and that NT4 may be useful in the therapy of acquired disorders of learning and memory.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 2000pg/ml,1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml human NT-4 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates or serum to each empty well. It is recommended that each human NT-4 standard solution and each sample be measured in duplicate.