Specifications
- Assay Type:Sandwich
- Reactivity:Mouse
- Description:Mouse NGF - NGF beta ELISA Kit
- Size:1 Kit
- Shipping temperature:Wet Ice
- Cat. no.:76236-758
Specifications
About this item
Mouse NGF - NGF beta AccuSignal ELISA Kit
- Natural and recombinant mouse NGF.
- There is no detectable cross-reactivity with other relevant proteins.
- Expression system for standard: E.coli; Immunogen sequence: S122-G241
Nerve growth factor(NGF) is a polypeptide involved in the regulation of growth and differentiation of sympathetic and certain sensory neurons. NGF is thought to have a profound effect on the development and maintenance of sympathetic and embryonic sensory neurones. NGF activity isolated from the male mouse submaxillary gland(MSG) consists of three types of subunits, alpha, beta and gamma, which specifically interact to form a 7S, approximately 130,000-molecular weight(Mr) complex. The 7S complex contains two identical 118-amino acid beta-chains, which are solely responsible for the nerve growth-stimulating activity of NGF. NGF, which is expressed by inflammatory cells and effects changes that lead to increased neural responsiveness, could be a pivotal mediator in allergic rhinitis.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 2000pg/ml, 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml mouse NGF standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernatants or serum to each empty well. It is recommended that each mouse NGF standard solution and each sample be measured in duplicate.