Specifications
- Reactivity:Human
- Target protein:MCP-1 (CCL2)
- Description:Human MCP-1 ELISA Kit
- Size:1 Kit
- Shipping temperature:Wet Ice
- Cat. no.:76236-712
Specifications
About this item
Human MCP-1 AccuSignal ELISA Kit
- Natural and recombinant human MCP-1.
- There is no detectable cross-reactivity with other relevant proteins.
- Expression system for standard: E.coli; Immunogen sequence: Q24-T99
Monocyte chemoattractant protein-1(MCP-1), a member of the chemokine(chemotactic cytokine) family, is a potent monocyte agonist that is upregulated by oxidized lipids. MCP-1 is also known as CCL2, SCYA2, MCAF. MCAF is a member of family of factors involved in immune and inflammatory responses. The amino acid sequence deduced from the nucleotide sequence reveals the primary structure of the MCAF precursor to be composed of a putative signal peptide sequence of 23 amino acid residues and a mature MCAF sequence of 76 amino acid residues. MCP-1 plays a unique and crucial role in the initiation of atherosclerosis and may provide a new therapeutic target in this disorder. Human MCP-1 is a 8.7KDa non-glycoprotein, consisting of 99 amino acids in precursor form and 76 amino acids in mature form.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml, 15.6pg/ml human MCP-1 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum, plasma(heparin, EDTA, citrate) or urine to each empty well. It is recommended that each human MCP-1 standard solution and each sample be measured in duplicate.