Specifications
- Reactivity:Human
- Target protein:Interleukin 10 (IL-10)
- Description:Human IL-10 ELISA Kit
- Size:1 Kit
- Shipping temperature:Wet Ice
- Cat. no.:76236-682
Specifications
About this item
Human Interleukin-10 AccuSignal ELISA Kit
- Natural and recombinant human IL-10.
- There is no detectable cross-reactivity with other relevant proteins.
- Expression system for standard: E.coli; Immunogen sequence: S19-N178
Interleukin-10, also called cytokine synthesis inhibitory factor, is implicated in tumorigenesis, and it has been shown that polymorphisms in its gene promoter correlate with differential amounts of production. IL-10 is an important cytokine with anti-inflammatory, anti-immune, and antifibrotic functions. It is also an important regulatory cytokine whose involvement extends into diverse areas of the human immune system. IL-10 is a recently described natural endogenous immunosuppressive cytokine that has been identified in human, murine, and other organisms. IL-10 significantly affects chemokine biology, because human IL-10 inhibits chemokine production and is a specific chemotactic factor for CD8+ T cells. It suppresses the ability of CD4+ T cells, but not CD8+ T cells, to migrate in response to IL-8. Interleukin-10 gene polymorphisms and interleukin-10 production capability may contribute to the development of skin squamous cell carcinomas after renal transplantation.1 The interleukin-10 locus contributes to the heritability of psoriasis susceptibility. With regard to sudden infant death, IL-10 is of special interest. This is an immunoregulatory cytokine that plays an important role in the development of infectious disease The mIL-10 gene is mapped to mouse chromosome 1 and the hIL-10 gene is also mapped to human chromosome 1. The standard product used in this kit is recombinant human IL-10, consisting of 160 amino acids with the molecular mass of 18.6KDa.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.3pg/ml, 15.6pg/ml, 7.8pg/ml human IL-10 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA, citrate) to each empty well. We recommend that each human IL-10 standard solution and each sample is measured in duplicate.