Specifications
- Reactivity:Human
- Target protein:IL-8 (CXCL8)
- Description:Human IL-8 ELISA Kit
- Size:1 Kit
- Shipping temperature:Wet Ice
- Cat. no.:76236-680
Specifications
About this item
Human Interleukin-8 AccuSignal ELISA Kit
- Natural and recombinant human IL-8.
- There is no detectable cross-reactivity with other relevant proteins.
- Expression system for standard: E.coli; Immunogen sequence: S28-S99
Interleukin-8, also called neutrophil-activating peptide-1 or SCYB8, is a tissue-derived peptide secreted by several types of cells in response to inflammatory stimuli. Monocyte-derived neutrophil chemotactic factor(MDNCF/IL-8, suggested gene symbol IL8) is a cytokine that chemoattracts and activates neutrophils. IL-8 is produced and released from human adipose tissue and from isolated adipocytes in vitro, which may indicate that IL-8 from adipose tissue could be involved in some of the obesity-related complications. The MDNCF/IL-8 gene is placed on the human gene map at position 4q12-q21. This is the same location where at least three other members(platelet factor 4, melanoma growth stimulatory activity, and interferon-gamma induced factor) of the platelet factor 4 gene superfamily reside. Human IL-8 consists of 99 amino acids in precursor form and 79 amino acids in mature form.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml, 15.6pg/ml human IL-8 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA, citrate) to each empty well. It is recommended that each human IL-8 standard solution and each sample be measured in duplicate.