Specifications
- Reactivity:Human
- Target protein:IL-6
- Description:Human IL-6 ELISA Kit
- Size:1 Kit
- Shipping temperature:Wet Ice
- Cat. no.:76236-674
Specifications
About this item
Human Interleukin-6 AccuSignal ELISA Kit
- Natural and recombinant human IL-6 .
- There is no detectable cross-reactivity with other relevant proteins.
- Expression system for standard: E.coli; Immunogen sequence: P29-M212
The human interferon-beta 2 gene(IFNB2) product is identical to that for the B-cell stimulation factor-2(BSF-2), the hybridoma growth factor(HGF)("interleukin-6"), and the hepatocyte stimulating factor(HSF). Proteins derived from this gene mediate the plasma protein response to tissue injury(acute-phase response) and regulate the growth and differentiation of both B and T cells. Interleukin-6(IL6) has come to be regarded as a potential osteoporotic factor because it has stimulatory effects on cells of the osteoclast lineage, and, thus, may play a role in the pathogenesis of bone loss associated with estrogen deficiency. IL-6 has many roles essential to the regulation of the immune response, hematopoiesis, and bone resorption. It is involved not only in the hepatic acute phase response but also in adipose tissue metabolism, lipoprotein lipase activity, and hepatic triglyceride secretion. Overproduction of IL-6, a proinflammatory cytokine, is associated with a spectrum of age-related conditions including cardiovascular disease, osteoporosis, arthritis, type 2 diabetes, certain cancers, periodontal disease, frailty, and functional decline. BSF-2 is a novel interleukin consisting of 184 amino acids. The standard product used in this kit is recombinant human IL-6, consisting of 184 amino acids with the molecular mass of 20.3kDa.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 300pg/ml, 150pg/ml, 75pg/ml, 37.5pg/ml, 18.75pg/ml, 9.38pg/ml, 4.69pg/ml human IL-6 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma( heparin, EDTA, citrate) to each empty well. We recommend that each human IL-6 standard solution and each sample is measured in duplicate.