Specifications
- Reactivity:Human
- Target protein:CD25 - IL-2sR alpha
- Description:Human CD25 - IL-2sR alpha ELISA Kit
- Size:1 Kit
- Shipping temperature:Wet Ice
- Cat. no.:76236-658
Specifications
About this item
Human CD25 - Interleukin-2sR alpha AccuSignal ELISA Kit
- Natural and recombinant human CD25.
- There is no detectable cross-reactivity with other relevant proteins.
- Expression system for standard: NSO; Immunogen sequence: E22-C213
CD25 is the alpha chain of the IL-2 receptor. It is a type I transmembrane protein present on activated T cells, activated B cells, some thymocytes, myeloid precursors, and oligodendrocytes that associates with CD122 to form a heterodimer that can act as a high-affinity receptor for IL-2. The IL2R gene was mapped to 10p15-p14. CD25 is expressed in most B-cell neoplasms, some acute nonlymphocytic leukemias, neuroblastomas, and tumor infiltrating lymphocytes. Its soluble form, called sIL-2R may be elevated in these diseases and is occasionally used to track disease progression.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 313pg/ml, 156pg/ml, 78pg/ml human CD25 standard solutions into the pre-coated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma( heparin, EDTA) to each empty well. We recommend that each human CD25 standard solution and each sample is measured in duplicate.