Order Entry
ContactUsLinkComponent
Rat ICAM-1 ELISA Kit, Rockland Immunochemicals
Rat ICAM-1 ELISA Kit, Rockland Immunochemicals
Catalog #: 76237-582
CAS Number:  
Rat ICAM-1 ELISA Kit, Rockland Immunochemicals
Catalog #: 76237-582
CAS Number:  

Specifications

  • Target protein:
    ICAM-1
  • Description:
    Rat ICAM-1 ELISA Kit
  • Size:
    1 Kit
  • Shipping temperature:
    Wet Ice
  • Cat. no.:
    76237-582

Specifications

About this item

Rat ICAM-1 AccuSignal ELISA Kit

  • Natural and recombinant rat ICAM-1.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: Q28-T493

Intercellular adhesion molecule-1(ICAM-1) is an integral membrane protein, a member of the immunoglobulin superfamily, and a ligand for lymphocyte function-associated(LFA) antigens, a beta 2 leukocyte integrin. The normal function of human intercellular adhesion molecule-1(ICAM-1) is to provide adhesion between endothelial cells and leukocytes after injury or stress. ICAM-1 binds to leukocyte function-associated antigen(LFA-1) or macrophage-1 antigen(Mac-1). ICAM-1 is found on leukocytes, fibroblasts, epithelial cells and endothelial cells and its expression is regulated by inflammatory cytokines. ICAM-1 has a tissue distribution similar to that of the major histocompatibility complex class II antigens and is likely to play a role in inflammatory responses. The standard product used in this kit is produced by recombinant gene expression, which is a dimeric protein linked with disulfide bonds.

Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 2000pg/ml, 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml rat ICAM-1 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of rat cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. We recommend that each rat ICAM-1 standard solution and each sample is measured in duplicate.

Customers Who Bought This Also Bought